Merck
  • Osteostatin Mitigates Gouty Arthritis through the Inhibition of Caspase-1 Activation and Upregulation of Nrf2 Expression.
Osteostatin Mitigates Gouty Arthritis through the Inhibition of Caspase-1 Activation and Upregulation of Nrf2 Expression.
International journal of molecular sciences (2024-03-13)
Laura Catalán, María Carmen Carceller, María Carmen Terencio, María José Alcaraz, María Luisa Ferrándiz, María Carmen Montesinos
ABSTRACT
Gouty arthritis results from monosodium urate (MSU) crystal deposition in joints, initiating (pro)-interleukin (IL)-1β maturation, inflammatory mediator release, and neutrophil infiltration, leading to joint swelling and pain. Parathyroid hormone-related protein (107-111) C-terminal peptide (osteostatin) has shown anti-inflammatory properties in osteoblasts and collagen-induced arthritis in mice, but its impact in gouty arthritis models remains unexplored. We investigated the effect of osteostatin on pyroptosis, inflammation, and oxidation in macrophages, as well as its role in the formation of calcium pyrophosphate dihydrate crystals and MSU-induced gouty arthritis in mice models. Osteostatin ameliorated pyroptosis induced by lipopolysaccharide and adenosine 5'-triphosphate (LPS + ATP) in mice peritoneal macrophages by reducing the expression of caspase-1, lactate dehydrogenase release, and IL-1β and IL-18 secretion. Additionally, IL-6 and tumor necrosis factor-α (TNF-α) were also decreased due to the reduced activation of the NF-κB pathway. Furthermore, osteostatin displayed antioxidant properties in LPS + ATP-stimulated macrophages, resulting in reduced production of mitochondrial and extracellular reactive oxygen species and enhanced Nrf2 translocation to the nuclei. In both models of gouty arthritis, osteostatin administration resulted in reduced pro-inflammatory cytokine production, decreased leukocyte migration, and reduced caspase-1 and NF-κB activation. These results highlight the potential of osteostatin as a therapeutic option for gouty arthritis.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution