Merck
  • Single substitution in H3.3G34 alters DNMT3A recruitment to cause progressive neurodegeneration.
Single substitution in H3.3G34 alters DNMT3A recruitment to cause progressive neurodegeneration.
Cell (2023-03-18)
Sima Khazaei, Carol C L Chen, Augusto Faria Andrade, Nisha Kabir, Pariya Azarafshar, Shahir M Morcos, Josiane Alves França, Mariana Lopes, Peder J Lund, Geoffroy Danieau, Samantha Worme, Lata Adnani, Nadine Nzirorera, Xiao Chen, Gayathri Yogarajah, Caterina Russo, Michele Zeinieh, Cassandra J Wong, Laura Bryant, Steven Hébert, Bethany Tong, Tianna S Sihota, Damien Faury, Evan Puligandla, Wajih Jawhar, Veronica Sandy, Mitra Cowan, Emily M Nakada, Loydie A Jerome-Majewska, Benjamin Ellezam, Carolina Cavalieri Gomes, Jonas Denecke, Davor Lessel, Marie T McDonald, Carolyn E Pizoli, Kathryn Taylor, Benjamin T Cocanougher, Elizabeth J Bhoj, Anne-Claude Gingras, Benjamin A Garcia, Chao Lu, Eric I Campos, Claudia L Kleinman, Livia Garzia, Nada Jabado
ABSTRACT
Germline histone H3.3 amino acid substitutions, including H3.3G34R/V, cause severe neurodevelopmental syndromes. To understand how these mutations impact brain development, we generated H3.3G34R/V/W knock-in mice and identified strikingly distinct developmental defects for each mutation. H3.3G34R-mutants exhibited progressive microcephaly and neurodegeneration, with abnormal accumulation of disease-associated microglia and concurrent neuronal depletion. G34R severely decreased H3K36me2 on the mutant H3.3 tail, impairing recruitment of DNA methyltransferase DNMT3A and its redistribution on chromatin. These changes were concurrent with sustained expression of complement and other innate immune genes possibly through loss of non-CG (CH) methylation and silencing of neuronal gene promoters through aberrant CG methylation. Complement expression in G34R brains may lead to neuroinflammation possibly accounting for progressive neurodegeneration. Our study reveals that H3.3G34-substitutions have differential impact on the epigenome, which underlie the diverse phenotypes observed, and uncovers potential roles for H3K36me2 and DNMT3A-dependent CH-methylation in modulating synaptic pruning and neuroinflammation in post-natal brains.
Sigma-Aldrich
Anti-OCT6 Antibody, clone KT110, clone KT110, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution