Merck
Selenium nanoparticles inhibit Staphylococcus aureus growth.
International journal of nanomedicine (2011-08-17)
Phong A Tran, Thomas J Webster
ABSTRACT
Staphylococcus aureus is a key bacterium commonly found in numerous infections. S. aureus infections are difficult to treat due to their biofilm formation and documented antibiotic resistance. While selenium has been used for a wide range of applications including anticancer applications, the effects of selenium nanoparticles on microorganisms remain largely unknown to date. The objective of this in vitro study was thus to examine the growth of S. aureus in the presence of selenium nanoparticles. Results of this study provided the first evidence of strongly inhibited growth of S. aureus in the presence of selenium nanoparticles after 3, 4, and 5 hours at 7.8, 15.5, and 31 μg/mL. The percentage of live bacteria also decreased in the presence of selenium nanoparticles. Therefore, this study suggests that selenium nanoparticles may be used to effectively prevent and treat S. aureus infections and thus should be further studied for such applications.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, crystallized, ≥98.0% (GE)
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)