- Structure of the human 20S U5 snRNP.
Structure of the human 20S U5 snRNP.
Nature structural & molecular biology (2024-03-12)
Sarah Schneider, Irina Brandina, Daniel Peter, Sonal Lagad, Angelique Fraudeau, Júlia Portell-Montserrat, Jonas Tholen, Jiangfeng Zhao, Wojciech P Galej
PMID38467877
ABSTRACT
The 20S U5 small nuclear ribonucleoprotein particle (snRNP) is a 17-subunit RNA-protein complex and a precursor of the U4/U6.U5 tri-snRNP, the major building block of the precatalytic spliceosome. CD2BP2 is a hallmark protein of the 20S U5 snRNP, absent from the mature tri-snRNP. Here we report a high-resolution cryogenic electron microscopy structure of the 20S U5 snRNP, shedding light on the mutually exclusive interfaces utilized during tri-snRNP assembly and the role of the CD2BP2 in facilitating this process.
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-2,2,7-Trimethylguanosine Mouse mAb (K121) Agarose Conjugate, suspension (Liquid), clone K121, Calbiochem®
Sigma-Aldrich
Anti-CD2BP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution