Merck
  • Combined bezafibrate and medroxyprogesterone acetate: potential novel therapy for acute myeloid leukaemia.
Combined bezafibrate and medroxyprogesterone acetate: potential novel therapy for acute myeloid leukaemia.
PloS one (2009-12-10)
Farhat L Khanim, Rachel E Hayden, Jane Birtwistle, Alessia Lodi, Stefano Tiziani, Nicholas J Davies, Jon P Ride, Mark R Viant, Ulrich L Gunther, Joanne C Mountford, Heinrich Schrewe, Richard M Green, Jim A Murray, Mark T Drayson, Chris M Bunce
ABSTRACT
The majority of acute myeloid leukaemia (AML) patients are over sixty years of age. With current treatment regimens, survival rates amongst these, and also those younger patients who relapse, remain dismal and novel therapies are urgently required. In particular, therapies that have anti-leukaemic activity but that, unlike conventional chemotherapy, do not impair normal haemopoiesis. Here we demonstrate the potent anti-leukaemic activity of the combination of the lipid-regulating drug bezafibrate (BEZ) and the sex hormone medroxyprogesterone acetate (MPA) against AML cell lines and primary AML cells. The combined activity of BEZ and MPA (B/M) converged upon the increased synthesis and reduced metabolism of prostaglandin D(2) (PGD(2)) resulting in elevated levels of the downstream highly bioactive, anti-neoplastic prostaglandin 15-deoxy Delta(12,14) PGJ(2) (15d-PGJ(2)). BEZ increased PGD(2) synthesis via the generation of reactive oxygen species (ROS) and activation of the lipid peroxidation pathway. MPA directed prostaglandin synthesis towards 15d-PGJ(2) by inhibiting the PGD(2) 11beta -ketoreductase activity of the aldo-keto reductase AKR1C3, which metabolises PGD(2) to 9alpha11beta-PGF(2alpha). B/M treatment resulted in growth arrest, apoptosis and cell differentiation in both AML cell lines and primary AML cells and these actions were recapitulated by treatment with 15d-PGJ(2). Importantly, the actions of B/M had little effect on the survival of normal adult myeloid progenitors. Collectively our data demonstrate that B/M treatment of AML cells elevated ROS and delivered the anti-neoplastic actions of 15d-PGJ(2). These observations provide the mechanistic rationale for the redeployment of B/M in elderly and relapsed AML.
Sigma-Aldrich
Ethanol, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology