Merck
  • Recapitulation of Skewed X-Inactivation in Female Ornithine Transcarbamylase-Deficient Primary Human Hepatocytes in the FRG Mouse: A Novel System for Developing Epigenetic Therapies.
Recapitulation of Skewed X-Inactivation in Female Ornithine Transcarbamylase-Deficient Primary Human Hepatocytes in the FRG Mouse: A Novel System for Developing Epigenetic Therapies.
Human gene therapy (2023-06-23)
Sharon C Cunningham, Eva B van Dijk, Erhua Zhu, Maya Sugden, Mawj Mandwie, Susan Siew, Beena Devanapalli, Adviye Ayper Tolun, Anne Klein, Laurence Wilson, Nader Aryamanesh, Paul Gissen, Julien Baruteau, Kaustuv Bhattacharya, Ian E Alexander
ABSTRACT
Realization of the immense therapeutic potential of epigenetic editing requires development of clinically predictive model systems that faithfully recapitulate relevant aspects of the target disease pathophysiology. In female patients with ornithine transcarbamylase (OTC) deficiency, an X-linked condition, skewed inactivation of the X chromosome carrying the wild-type OTC allele is associated with increased disease severity. The majority of affected female patients can be managed medically, but a proportion require liver transplantation. With rapid development of epigenetic editing technology, reactivation of silenced wild-type OTC alleles is becoming an increasingly plausible therapeutic approach. Toward this end, privileged access to explanted diseased livers from two affected female infants provided the opportunity to explore whether engraftment and expansion of dissociated patient-derived hepatocytes in the FRG mouse might produce a relevant model for evaluation of epigenetic interventions. Hepatocytes from both infants were successfully used to generate chimeric mouse-human livers, in which clusters of primary human hepatocytes were either OTC positive or negative by immunohistochemistry (IHC), consistent with clonal expansion from individual hepatocytes in which the mutant or wild-type OTC allele was inactivated, respectively. Enumeration of the proportion of OTC-positive or -negative human hepatocyte clusters was consistent with dramatic skewing in one infant and minimal to modest skewing in the other. Importantly, IHC and fluorescence-activated cell sorting analysis of intact and dissociated liver samples from both infants showed qualitatively similar patterns, confirming that the chimeric mouse-human liver model recapitulated the native state in each infant. Also of importance was the induction of a treatable metabolic phenotype, orotic aciduria, in mice, which correlated with the presence of clonally expanded OTC-negative primary human hepatocytes. We are currently using this unique model to explore CRISPR-dCas9-based epigenetic targeting strategies in combination with efficient adeno-associated virus (AAV) gene delivery to reactivate the silenced functional OTC gene on the inactive X chromosome.
Sigma-Aldrich
Anti-OTC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution