Merck
  • Pharmacokinetic-pharmacodynamic modelling of fluvoxamine 5-HT transporter occupancy in rat frontal cortex.
Pharmacokinetic-pharmacodynamic modelling of fluvoxamine 5-HT transporter occupancy in rat frontal cortex.
British journal of pharmacology (2008-05-22)
M Geldof, J I Freijer, L van Beijsterveldt, X Langlois, M Danhof
ABSTRACT
The pharmacokinetic-pharmacodynamic (PK-PD) correlation of fluvoxamine 5-HT transporter (SERT) occupancy was determined in rat frontal cortex ex vivo. Rats (n=47) with permanent arterial and venous cannulas received a 30 min intravenous infusion of fluvoxamine (1 or 7.3 mg kg(-1)). At various time points after dosing, brains were collected for determination of fluvoxamine concentration and SERT occupancy. In addition, the time course of fluvoxamine concentration in plasma was determined up to the time of brain collection. In a separate study (n=26), the time course of fluvoxamine concentration in brain extracellular fluid (ECF) and plasma was determined. The results of the investigations were interpreted by nonlinear mixed effects modeling. Highest SERT occupancy was reached at the first time point (10 or 15 min) and maintained for 1.5 and 7 h after 1 and 7.3 mg kg(-1), respectively. Thereafter, SERT occupancy decreased linearly at a rate of 8% h(-1). SERT occupancy could be directly related to plasma, brain ECF and brain tissue concentrations by a hyperbolic function (Bmax model). Maximal SERT occupancy (Bmax) was 95%. Estimated concentrations at half-maximal SERT occupancy (EC50) in plasma, ECF and brain tissue were 0.48, 0.22 and 14.8 ng mL(-1) respectively. The minimum value of the objective function decreased 12 points for ECF and brain tissue concentrations relative to plasma (P<0.01), presumably as a result of nonlinear brain distribution. The proposed PK-PD model constitutes a useful basis for prediction of the time course of ex vivo SERT occupancy in behavioural studies with selective serotonin reuptake inhibitors.
Product Description
Supelco
Heptane, analytical standard
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, lyophilized powder, essentially fatty acid free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated