Merck
  • In infertile women, cells from Chlamydia trachomatis infected sites release higher levels of interferon-gamma, interleukin-10 and tumor necrosis factor-alpha upon heat-shock-protein stimulation than fertile women.
In infertile women, cells from Chlamydia trachomatis infected sites release higher levels of interferon-gamma, interleukin-10 and tumor necrosis factor-alpha upon heat-shock-protein stimulation than fertile women.
Reproductive biology and endocrinology : RB&E (2008-05-21)
Pragya Srivastava, Rajneesh Jha, Sylvette Bas, Sudha Salhan, Aruna Mittal
ABSTRACT
The magnitude of reproductive morbidity associated with sexually transmitted Chlamydia trachomatis infection is enormous. Association of antibodies to chlamydial heat shock proteins (cHSP) 60 and 10 with various disease sequelae such as infertility or ectopic pregnancy has been reported. Cell-mediated immunity is essential in resolution and in protection to Chlamydia as well as is involved in the immunopathogenesis of chlamydial diseases. To date only peripheral cell mediated immune responses have been evaluated for cHSP60. These studies suggest cHSPs as important factors involved in immunopathological condition associated with infection. Hence study of specific cytokine responses of mononuclear cells from the infectious site to cHSP60 and cHSP10 may elucidate their actual role in the cause of immunopathogenesis and the disease outcome. Female patients (n = 368) attending the gynecology out patient department of Safdarjung hospital, New Delhi were enrolled for the study and were clinically characterized into two groups; chlamydia positive fertile women (n = 63) and chlamydia positive infertile women (n = 70). Uninfected healthy women with no infertility problem were enrolled as controls (n = 39). cHSP60 and cHSP10 specific cytokine responses (Interferon (IFN)-gamma, Interleukin (IL)-10, Tumor Necrosis Factor (TNF)-alpha, IL-13 and IL-4) were assessed by ELISA in stimulated cervical mononuclear cell supernatants. cHSP60 and cHSP10 stimulation results in significant increase in IFN-gamma (P = 0.006 and P = 0.04 respectively) and IL-10 levels (P = 0.04) in infertile group as compared to fertile group. A significant cHSP60 specific increase in TNF-alpha levels (P = 0.0008) was observed in infertile group as compared to fertile group. cHSP60 and cHSP10 specific IFN-gamma and IL-10 levels were significantly correlated (P < 0.0001, r = 0.54 and P = 0.004, r = 0.33 respectively) in infertile group. Our results suggest that exposure to chlamydial heat shock proteins (cHSP60 and cHSP10) could significantly affect mucosal immune function by increasing the release of IFN-gamma, IL-10 and TNF-alpha by cervical mononuclear cells.
Sigma-Aldrich
Polymyxin B-Agarose, aqueous glycerol suspension
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
SAFC
RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
Sigma-Aldrich
RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
SAFC
RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications