Merck
  • The mechanosensitive TRPV2 calcium channel promotes human melanoma invasiveness and metastatic potential.
The mechanosensitive TRPV2 calcium channel promotes human melanoma invasiveness and metastatic potential.
EMBO reports (2023-02-07)
Kenji F Shoji, Elsa Bayet, Sabrina Leverrier-Penna, Dahiana Le Devedec, Aude Mallavialle, Séverine Marionneau-Lambot, Florian Rambow, Raul Perret, Aurélie Joussaume, Roselyne Viel, Alain Fautrel, Amir Khammari, Bruno Constantin, Sophie Tartare-Deckert, Aubin Penna
ABSTRACT
Melanoma is a highly aggressive cancer endowed with a unique capacity of rapidly metastasizing, which is fundamentally driven by aberrant cell motility behaviors. Discovering "migrastatics" targets, specifically controlling invasion and dissemination of melanoma cells during metastasis, is therefore of primary importance. Here, we uncover the prominent expression of the plasma membrane TRPV2 calcium channel as a distinctive feature of melanoma tumors, directly related to melanoma metastatic dissemination. In vitro as well as in vivo, TRPV2 activity is sufficient to confer both migratory and invasive potentials, while conversely TRPV2 silencing in highly metastatic melanoma cells prevents aggressive behavior. In invasive melanoma cells, TRPV2 channel localizes at the leading edge, in dynamic nascent adhesions, and regulates calcium-mediated activation of calpain and the ensuing cleavage of the adhesive protein talin, along with F-actin organization. In human melanoma tissues, TRPV2 overexpression correlates with advanced malignancy and poor prognosis, evoking a biomarker potential. Hence, by regulating adhesion and motility, the mechanosensitive TRPV2 channel controls melanoma cell invasiveness, highlighting a new therapeutic option for migrastatics in the treatment of metastatic melanoma.
Sigma-Aldrich
Anti-TRPV2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Tranilast, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Talin antibody produced in mouse, clone 8d4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid