Merck
  • Intermedin1‑47 inhibits high phosphate‑induced vascular smooth muscle cell calcification by regulating Wnt/β‑catenin signaling.
Intermedin1‑47 inhibits high phosphate‑induced vascular smooth muscle cell calcification by regulating Wnt/β‑catenin signaling.
Molecular medicine reports (2021-08-21)
Yin Zhang, Naiwang Tang, Jinjie Zhou
ABSTRACT
Vascular calcification is a major risk factor for cardiovascular disease and accounts for a large proportion of deaths from cardiovascular disease in patients with chronic kidney disease. The high incidence, rapid progression and irreversibility of vascular smooth muscle cell (VSMC) calcification in patients has attracted attention. In the present study, the effect of intermedin1‑47 (IMD1‑47), an important isoform of intermedin, was investigated on the calcification of rat cardiovascular VSMCs induced by high phosphate (HP). To stimulate osteoblast‑like differentiation and calcification in rat VSMCs, 10 mM β‑sodium glycerophosphate was used. The VSMCs were then treated with three doses of IMD1‑47 and the effects of IMD1‑47 on VSMC calcification, on the expression of osteogenic markers [osteoprotegerin, Runt‑related transcription factor 2 (Runx2) and osteopontin (OPN)] and on alkaline phosphatase (ALP) activity were assessed. HP treatment significantly enhanced the cellular calcium content of VSMCs, the expression of osteogenic markers, and ALP activity, while IMD1‑47 significantly reversed these effects in a dose‑dependent manner. The protein expression levels of Wnt1, Wnt3a and active β‑catenin were determined and it was found that IMD1‑47 significantly inhibited their expression. Following β‑catenin silencing, the protein expression levels Runx2 and OPN were increased compared with the IMD1‑47 treatment alone, indicating a role for the Wnt/β‑catenin pathway in the effects of IMD1‑47 on osteogenic markers. The present study suggested that IMD1‑47 inhibited HP‑induced VSMC calcification by regulating the Wnt/β‑catenin signaling pathway.
Sigma-Aldrich
ANTI-OPN-A/B(N-TERMINAL) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-WNT3A antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Osteoprotegerin antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-WNT1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
ANTI-RUNX2 antibody produced in mouse, clone 4E5, ascites fluid
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse