Merck
  • Loss of non-motor kinesin KIF26A causes congenital brain malformations via dysregulated neuronal migration and axonal growth as well as apoptosis.
Loss of non-motor kinesin KIF26A causes congenital brain malformations via dysregulated neuronal migration and axonal growth as well as apoptosis.
Developmental cell (2022-10-14)
Xuyu Qian, Ellen M DeGennaro, Maya Talukdar, Shyam K Akula, Abbe Lai, Diane D Shao, Dilenny Gonzalez, Jack H Marciano, Richard S Smith, Norma K Hylton, Edward Yang, J Fernando Bazan, Lee Barrett, Rebecca C Yeh, R Sean Hill, Samantha G Beck, Aoi Otani, Jolly Angad, Tadahiro Mitani, Jennifer E Posey, Davut Pehlivan, Daniel Calame, Hatip Aydin, Osman Yesilbas, Kendall C Parks, Emanuela Argilli, Eleina England, Kiho Im, Ajay Taranath, Hamish S Scott, Christopher P Barnett, Peer Arts, Elliott H Sherr, James R Lupski, Christopher A Walsh
ABSTRACT
Kinesins are canonical molecular motors but can also function as modulators of intracellular signaling. KIF26A, an unconventional kinesin that lacks motor activity, inhibits growth-factor-receptor-bound protein 2 (GRB2)- and focal adhesion kinase (FAK)-dependent signal transduction, but its functions in the brain have not been characterized. We report a patient cohort with biallelic loss-of-function variants in KIF26A, exhibiting a spectrum of congenital brain malformations. In the developing brain, KIF26A is preferentially expressed during early- and mid-gestation in excitatory neurons. Combining mice and human iPSC-derived organoid models, we discovered that loss of KIF26A causes excitatory neuron-specific defects in radial migration, localization, dendritic and axonal growth, and apoptosis, offering a convincing explanation of the disease etiology in patients. Single-cell RNA sequencing in KIF26A knockout organoids revealed transcriptional changes in MAPK, MYC, and E2F pathways. Our findings illustrate the pathogenesis of KIF26A loss-of-function variants and identify the surprising versatility of this non-motor kinesin.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-KIF26A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution