Merck
  • Generation of locus coeruleus norepinephrine neurons from human pluripotent stem cells.
Generation of locus coeruleus norepinephrine neurons from human pluripotent stem cells.
Nature biotechnology (2023-11-17)
Yunlong Tao, Xueyan Li, Qiping Dong, Linghai Kong, Andrew J Petersen, Yuanwei Yan, Ke Xu, Seth Zima, Yanru Li, Danielle K Schmidt, Melvin Ayala, Sakthikumar Mathivanan, Andre M M Sousa, Qiang Chang, Su-Chun Zhang
ABSTRACT
Central norepinephrine (NE) neurons, located mainly in the locus coeruleus (LC), are implicated in diverse psychiatric and neurodegenerative diseases and are an emerging target for drug discovery. To facilitate their study, we developed a method to generate 40-60% human LC-NE neurons from human pluripotent stem cells. The approach depends on our identification of ACTIVIN A in regulating LC-NE transcription factors in dorsal rhombomere 1 (r1) progenitors. In vitro generated human LC-NE neurons display extensive axonal arborization; release and uptake NE; and exhibit pacemaker activity, calcium oscillation and chemoreceptor activity in response to CO2. Single-nucleus RNA sequencing (snRNA-seq) analysis at multiple timepoints confirmed NE cell identity and revealed the differentiation trajectory from hindbrain progenitors to NE neurons via an ASCL1-expressing precursor stage. LC-NE neurons engineered with an NE sensor reliably reported extracellular levels of NE. The availability of functional human LC-NE neurons enables investigation of their roles in psychiatric and neurodegenerative diseases and provides a tool for therapeutics development.
Sigma-Aldrich
Strychnine hydrochloride
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder
Sigma-Aldrich
(+)-Bicuculline, ≥97.0% (TLC)
Sigma-Aldrich
D(−)-2-Amino-5-phosphonopentanoic acid, NMDA receptor antagonist
Sigma-Aldrich
Anti-Adrenergic Receptor α-2A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
CNQX disodium salt hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-GAL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution