Merck
  • Ciliated Cells in Ovarian Cancer Decrease with Increasing Tumor Grade and Disease Progression.
Ciliated Cells in Ovarian Cancer Decrease with Increasing Tumor Grade and Disease Progression.
Cells (2022-12-24)
Michael T Richardson, Maria Sol Recouvreux, Beth Y Karlan, Ann E Walts, Sandra Orsulic
ABSTRACT
Ciliated cell markers expressed in epithelial ovarian cancers (EOC) are associated with improved survival. We examined the distribution of cells expressing ciliated cell markers in various EOC histologies and stages. Immunohistochemistry and/or multiplex immunofluorescence were used to determine the expression of FOXJ1 and/or CAPS (ciliated cell markers) in tissue microarrays including 4 normal fallopian tubes, 6 normal endometria, 16 cystadenomas, 25 borderline tumors, 21 low-grade carcinomas, and 118 high-grade carcinomas (HGSOC) (46 serous, 29 endometrioid, 30 clear cell, 13 mucinous). CAPS+ cells were observed in normal fallopian tubes and endometria and in ~85% of serous benign and borderline tumors and low-grade carcinomas but only in <40% of HGSOC. mRNA data from an independent cohort showed higher FOXJ1 and CAPS expression in serous borderline tumors and low-grade carcinomas compared to HGSOC. In HGSOC, ciliated cell-positive markers were observed in 52% primary tumors compared to 26% of patient-matched synchronous metastases, and 24% metachronous metastases (p = 0.009). mRNA data from an independent HGSOC cohort showed lower levels of CAPS in metastases than in primary tumors (p = 0.03). Overall, the study revealed that ciliated cells were less common in mucinous EOC, the percentage of ciliated cell marker-positive cases decreased with increasing grade, and the percentage of ciliated cells decreased in HGSOC metastases compared to patient-matched primary tumors.
Sigma-Aldrich
Anti-FOXJ1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-CAPS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution