Merck
  • Endogenous Inflammatory Mediators Produced by Injury Activate TRPV1 and TRPA1 Nociceptors to Induce Sexually Dimorphic Cold Pain That Is Dependent on TRPM8 and GFRα3.
Endogenous Inflammatory Mediators Produced by Injury Activate TRPV1 and TRPA1 Nociceptors to Induce Sexually Dimorphic Cold Pain That Is Dependent on TRPM8 and GFRα3.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2023-03-11)
Chenyu Yang, Shanni Yamaki, Tyler Jung, Brian Kim, Ryan Huyhn, David D McKemy
ABSTRACT
The detection of environmental temperatures is critical for survival, yet inappropriate responses to thermal stimuli can have a negative impact on overall health. The physiological effect of cold is distinct among somatosensory modalities in that it is soothing and analgesic, but also agonizing in the context of tissue damage. Inflammatory mediators produced during injury activate nociceptors to release neuropeptides, such as calcitonin gene-related peptide (CGRP) and substance P, inducing neurogenic inflammation, which further exasperates pain. Many inflammatory mediators induce sensitization to heat and mechanical stimuli but, conversely, inhibit cold responsiveness, and the identity of molecules inducing cold pain peripherally is enigmatic, as are the cellular and molecular mechanisms altering cold sensitivity. Here, we asked whether inflammatory mediators that induce neurogenic inflammation via the nociceptive ion channels TRPV1 (vanilloid subfamily of transient receptor potential channel) and TRPA1 (transient receptor potential ankyrin 1) lead to cold pain in mice. Specifically, we tested cold sensitivity in mice after intraplantar injection of lysophosphatidic acid or 4-hydroxy-2-nonenal, finding that each induces cold pain that is dependent on the cold-gated channel transient receptor potential melastatin 8 (TRPM8). Inhibition of CGRP, substance P, or toll-like receptor 4 (TLR4) signaling attenuates this phenotype, and each neuropeptide produces TRPM8-dependent cold pain directly. Further, the inhibition of CGRP or TLR4 signaling alleviates cold allodynia differentially by sex. Last, cold pain induced by both inflammatory mediators and neuropeptides requires TRPM8, as well as the neurotrophin artemin and its receptor GDNF receptor α3 (GFRα3). These results are consistent with artemin-induced cold allodynia requiring TRPM8, demonstrating that neurogenic inflammation alters cold sensitivity via localized artemin release that induces cold pain via GFRα3 and TRPM8.SIGNIFICANCE STATEMENT The cellular and molecular mechanisms that generate pain are complex with a diverse array of pain-producing molecules generated during injury that act to sensitize peripheral sensory neurons, thereby inducing pain. Here we identify a specific neuroinflammatory pathway involving the ion channel TRPM8 (transient receptor potential cation channel subfamily M member 8) and the neurotrophin receptor GFRα3 (GDNF receptor α3) that leads to cold pain, providing select targets for potential therapies for this pain modality.
Sigma-Aldrich
TLR4 Inhibitor, TAK-242, TAK-242; CAS 243984-11-4; is a cell-permeable compound that selectively binds to Cys747 of TLR4 and selectively disrupts its interaction with adaptor molecules TIRAP and TRAM.