Merck
  • Activation of an actin signaling pathway in pre-malignant mammary epithelial cells by P-cadherin is essential for transformation.
Activation of an actin signaling pathway in pre-malignant mammary epithelial cells by P-cadherin is essential for transformation.
Disease models & mechanisms (2023-02-23)
Lídia Faria, Sara Canato, Tito T Jesus, Margarida Gonçalves, Patrícia S Guerreiro, Carla S Lopes, Isabel Meireles, Eurico Morais-de-Sá, Joana Paredes, Florence Janody
ABSTRACT
Alterations in the expression or function of cell adhesion molecules have been implicated in all steps of tumor progression. Among those, P-cadherin is highly enriched in basal-like breast carcinomas, playing a central role in cancer cell self-renewal, collective cell migration and invasion. To establish a clinically relevant platform for functional exploration of P-cadherin effectors in vivo, we generated a humanized P-cadherin Drosophila model. We report that actin nucleators, Mrtf and Srf, are main P-cadherin effectors in fly. We validated these findings in a human mammary epithelial cell line with conditional activation of the SRC oncogene. We show that, prior to promoting malignant phenotypes, SRC induces a transient increase in P-cadherin expression, which correlates with MRTF-A accumulation, its nuclear translocation and the upregulation of SRF target genes. Moreover, knocking down P-cadherin, or preventing F-actin polymerization, impairs SRF transcriptional activity. Furthermore, blocking MRTF-A nuclear translocation hampers proliferation, self-renewal and invasion. Thus, in addition to sustaining malignant phenotypes, P-cadherin can also play a major role in the early stages of breast carcinogenesis by promoting a transient boost of MRTF-A-SRF signaling through actin regulation.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
CCG-203971, ≥98% (HPLC)