Merck
  • Host cell egress of Brucella abortus requires BNIP3L-mediated mitophagy.
Host cell egress of Brucella abortus requires BNIP3L-mediated mitophagy.
The EMBO journal (2023-05-26)
Jérémy Verbeke, Youri Fayt, Lisa Martin, Oya Yilmaz, Jaroslaw Sedzicki, Angéline Reboul, Michel Jadot, Patricia Renard, Christoph Dehio, Henri-François Renard, Jean-Jacques Letesson, Xavier De Bolle, Thierry Arnould
ABSTRACT
The facultative intracellular pathogen Brucella abortus interacts with several organelles of the host cell to reach its replicative niche inside the endoplasmic reticulum. However, little is known about the interplay between the intracellular bacteria and the host cell mitochondria. Here, we showed that B. abortus triggers substantive mitochondrial network fragmentation, accompanied by mitophagy and the formation of mitochondrial Brucella-containing vacuoles during the late steps of cellular infection. Brucella-induced expression of the mitophagy receptor BNIP3L is essential for these events and relies on the iron-dependent stabilisation of the hypoxia-inducible factor 1α. Functionally, BNIP3L-mediated mitophagy appears to be advantageous for bacterial exit from the host cell as BNIP3L depletion drastically reduces the number of reinfection events. Altogether, these findings highlight the intricate link between Brucella trafficking and the mitochondria during host cell infection.
Sigma-Aldrich
Anti-FAM92A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Anti-EF5 Antibody, clone ELK3-51 Antibody, Cyanine 5 conjugate, clone ELK3-51, from mouse, CY5 conjugate
Sigma-Aldrich
Anti-LC3B antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid