Merck
  • Pepsin-digested chicken-liver hydrolysate attenuates hepatosteatosis by relieving hepatic and peripheral insulin resistance in long-term high-fat dietary habit.
Pepsin-digested chicken-liver hydrolysate attenuates hepatosteatosis by relieving hepatic and peripheral insulin resistance in long-term high-fat dietary habit.
Journal of food and drug analysis (2022-06-14)
Yi-Hsieng Samuel Wu, Yi-Ling Lin, Wen-Yuan Yang, Sheng-Yao Wang, Yi-Chen Chen
ABSTRACT
This study aims to clarify the effects of chicken liver hydrolysates (CLHs) on long-term high-fat diet (HFD)-induced insulin resistance (IR) and hepatosteatosis in mice. In vitro, the 400 μM oleic acid (OA)-added medium successfully stimulated the cellular steatosis on FL83B cells, and the cellular steatosis was attenuated ( p < 0.05) by supplementing with CLHs (4 mg/L). In vivo, the effects of CLHs on IR and hepatosteatosis development were tested in 20-week HFD-fed mice. HFD-induced increases in final body weight, but body weight gains of mice were decreased ( p < 0.05) by supplementing CLHs. Elevated ( p < 0.05) serum aspartate aminotransferase (AST), alanine transaminase (ALT), alkaline phosphatase (ALP), free fatty acids (FFAs), triglyceride (TG), total cholesterol (TC), and fasted glucose values in HFD-fed mice decreased ( p < 0.05) by supplementing CLHs. Both results of hepatic steatosis and fibrotic scores also indicated the retardation ( p < 0.05) of the hepatosteatosis in cotreated groups. Moreover, the CLH supplementation sustained ( p < 0.05) hepatic and peripheral insulin signal sensitivity in HFD-fed mice. CLH supplementation could ameliorate hepatic lipid deposition, hepatic/peripheral IR in a long-term high-fat dietary habit, and also improve the universal glucose homeostasis by upregulating hepatic and peripheral insulin sensitivities.
Sigma-Aldrich
Anti-CPT1A Antibody, serum, from rabbit
Sigma-Aldrich
Anti-Uncoupling Protein-2 (UCP-2) (144-157) Rabbit pAb, liquid, Calbiochem®