MilliporeSigma
Vas deferens calculus.
The Journal of urology (1989-02-01)
S Küpeli, M Safak, L S Yaman
ABSTRACT
We describe a patient with a radiopaque calculus in the vas deferens that was inappropriately diagnosed as a ureteral calculus.
Sigma-Aldrich
Vatiquinone, ≥95% (HPLC)