Merck
  • Mapping PTBP2 binding in human brain identifies SYNGAP1 as a target for therapeutic splice switching.
Mapping PTBP2 binding in human brain identifies SYNGAP1 as a target for therapeutic splice switching.
Nature communications (2023-05-07)
Jennine M Dawicki-McKenna, Alex J Felix, Elisa A Waxman, Congsheng Cheng, Defne A Amado, Paul T Ranum, Alexey Bogush, Lea V Dungan, Jean Ann Maguire, Alyssa L Gagne, Elizabeth A Heller, Deborah L French, Beverly L Davidson, Benjamin L Prosser
ABSTRACT
Alternative splicing of neuronal genes is controlled partly by the coordinated action of polypyrimidine tract binding proteins (PTBPs). While PTBP1 is ubiquitously expressed, PTBP2 is predominantly neuronal. Here, we define the PTBP2 footprint in the human transcriptome using brain tissue and human induced pluripotent stem cell-derived neurons (iPSC-neurons). We map PTBP2 binding sites, characterize PTBP2-dependent alternative splicing events, and identify novel PTBP2 targets including SYNGAP1, a synaptic gene whose loss-of-function leads to a complex neurodevelopmental disorder. We find that PTBP2 binding to SYNGAP1 mRNA promotes alternative splicing and nonsense-mediated decay, and that antisense oligonucleotides (ASOs) that disrupt PTBP binding redirect splicing and increase SYNGAP1 mRNA and protein expression. In SYNGAP1 haploinsufficient iPSC-neurons generated from two patients, we show that PTBP2-targeting ASOs partially restore SYNGAP1 expression. Our data comprehensively map PTBP2-dependent alternative splicing in human neurons and cerebral cortex, guiding development of novel therapeutic tools to benefit neurodevelopmental disorders.
Sigma-Aldrich
Anti-PTBP2 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-MAP2 (2a+2b) antibody, Mouse monoclonal, clone AP-20, ascites fluid
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)