- PEG2000-DBCO surface coating increases intracellular uptake of liposomes by breast cancer xenografts.
PEG2000-DBCO surface coating increases intracellular uptake of liposomes by breast cancer xenografts.
Given our interest in the utility of liposomes for molecular imaging and theranostics, we investigated how coating the outer layer of the liposome affects internalization by breast cancer cell lines in vitro and in breast tumor tissues in vivo. Indeed, we discovered that a remarkably high liposomal uptake can be achieved by DBCO (dibenzocyclooctyne) soft coating. Our data demonstrates that decorating the terminal lipid with a DBCO moiety at a specific density induces increased tumor uptake in vivo (tumor uptake ~ 50%) compared to conventional undecorated liposome (tumor uptake ~ 20%). In this study, we report improved visualization of breast cancer cells in vivo using a 4T1 orthotopic breast cancer model and primary breast tumor xenograft models MDA-MB-231 and MDA-MB-436. L-PEG2000-DBCO coated liposomes demonstrate increased accumulation in breast cancer cells independent of tumor size, type, position, receptor expression, as well as the condition of the host mice. We expect these findings to have a major positive impact on the practical utility of liposomes in image-guided applications and precision medicine theranostics.