Merck
  • The endoplasmic reticulum kinase PERK interacts with the oxidoreductase ERO1 to metabolically adapt mitochondria.
The endoplasmic reticulum kinase PERK interacts with the oxidoreductase ERO1 to metabolically adapt mitochondria.
Cell reports (2023-01-01)
Arthur Bassot, Junsheng Chen, Kei Takahashi-Yamashiro, Megan C Yap, Christine Silvia Gibhardt, Giang N T Le, Saaya Hario, Yusuke Nasu, Jack Moore, Tomas Gutiérrez, Lucas Mina, Heather Mast, Audric Moses, Rakesh Bhat, Klaus Ballanyi, Hélène Lemieux, Roberto Sitia, Ester Zito, Ivan Bogeski, Robert E Campbell, Thomas Simmen
ABSTRACT
Endoplasmic reticulum (ER) homeostasis requires molecular regulators that tailor mitochondrial bioenergetics to the needs of protein folding. For instance, calnexin maintains mitochondria metabolism and mitochondria-ER contacts (MERCs) through reactive oxygen species (ROS) from NADPH oxidase 4 (NOX4). However, induction of ER stress requires a quick molecular rewiring of mitochondria to adapt to new energy needs. This machinery is not characterized. We now show that the oxidoreductase ERO1⍺ covalently interacts with protein kinase RNA-like ER kinase (PERK) upon treatment with tunicamycin. The PERK-ERO1⍺ interaction requires the C-terminal active site of ERO1⍺ and cysteine 216 of PERK. Moreover, we show that the PERK-ERO1⍺ complex promotes oxidization of MERC proteins and controls mitochondrial dynamics. Using proteinaceous probes, we determined that these functions improve ER-mitochondria Ca2+ flux to maintain bioenergetics in both organelles, while limiting oxidative stress. Therefore, the PERK-ERO1⍺ complex is a key molecular machinery that allows quick metabolic adaptation to ER stress.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
ISRIB, ≥98% (HPLC)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Anti-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Tunicamycin, Streptomyces lysosuperficus
Sigma-Aldrich
Anti-Ero1 Antibody, clone 2G4/12, clone 2G4/12, from mouse
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody