Merck
  • Essential role of Rpd3-dependent lysine modification in the growth, development and virulence of Beauveria bassiana.
Essential role of Rpd3-dependent lysine modification in the growth, development and virulence of Beauveria bassiana.
Environmental microbiology (2018-03-27)
Qing Cai, Zhi-Kang Wang, Wei Shao, Sheng-Hua Ying, Ming-Guang Feng
ABSTRACT
Rpd3 is a class I histone deacetylase that reverses lysine acetylation thus influencing cellular processes and functions. However, its role in fungal insect pathogens has not been explored yet. Here we show that Rpd3-dependent lysine modification and gene expression orchestrate growth, conidiation and virulence in Beauveria bassiana. Deletion of Rpd3 resulted in severe growth defects on various carbon/nitrogen sources, 97% reduction in conidiation capacity and drastic attenuation in virulence. These phenotypes concurred with differential expression of 1479 proteins and hyperacetylation or hypoacetylation of 2227 lysine residues on 1134 proteins. Many of these proteins fell into carbon/nitrogen metabolism and cell rescue/defence/virulence, indicating vital roles of Rpd3-dependent protein expression and lysine modification in the fungal growth and virulence. Intriguingly, lysine residues of four core histones (H2A, H2B, H3 and H4) and many histone acetyltransferases were also hyper- or hypoacetylated in Δrpd3, suggesting direct and indirect roles for Rpd3 in genome-wide lysine modification. However, crucial development activators were transcriptionally repressed and not found in either proteome or acetylome. Single/double-site-directed H3K9/K14 mutations for hyper/hypoacetylation exerted significant impacts on conidiation and dimorphic transition crucial for fungal virulence. Altogether, Rpd3 mediates growth, asexual development and virulence through transcriptional/translational regulation and posttranslational lysine modification in B. bassiana.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys14) Antibody, serum, Upstate®
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys16) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys56) Antibody, from rabbit, purified by affinity chromatography