Merck
  • Mitochondrial Transfer Induced by Adipose-Derived Mesenchymal Stem Cell Transplantation Improves Cardiac Function in Rat Models of Ischemic Cardiomyopathy.
Mitochondrial Transfer Induced by Adipose-Derived Mesenchymal Stem Cell Transplantation Improves Cardiac Function in Rat Models of Ischemic Cardiomyopathy.
Cell transplantation (2023-01-11)
Daisuke Mori, Shigeru Miyagawa, Takuji Kawamura, Daisuke Yoshioka, Hiroki Hata, Takayoshi Ueno, Koichi Toda, Toru Kuratani, Miwa Oota, Kotoe Kawai, Hayato Kurata, Hiroyuki Nishida, Akima Harada, Toshihiko Toyofuku, Yoshiki Sawa
ABSTRACT
Although mesenchymal stem cell transplantation has been successful in the treatment of ischemic cardiomyopathy, the underlying mechanisms remain unclear. Herein, we investigated whether mitochondrial transfer could explain the success of cell therapy in ischemic cardiomyopathy. Mitochondrial transfer in co-cultures of human adipose-derived mesenchymal stem cells and rat cardiomyocytes maintained under hypoxic conditions was examined. Functional recovery was monitored in a rat model of myocardial infarction following human adipose-derived mesenchymal stem cell transplantation. We observed mitochondrial transfer in vitro, which required the formation of cell-to-cell contacts and synergistically enhanced energy metabolism. Rat cardiomyocytes exhibited mitochondrial transfer 3 days following human adipose-derived mesenchymal stem cell transplantation to the ischemic heart surface post-myocardial infarction. We detected donor mitochondrial DNA in the recipient myocardium concomitant with a significant improvement in cardiac function. Mitochondrial transfer is vital for successful cell transplantation therapies and improves treatment outcomes in ischemic cardiomyopathy.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
ANTI-VIMENTIN (S82) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody (rabbit), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-von Willebrand Factor Antibody, Chemicon®, from rabbit