MilliporeSigma
  • Double knockin mice show NF-κB trajectories in immune signaling and aging.
Double knockin mice show NF-κB trajectories in immune signaling and aging.
Cell reports (2022-11-24)
Shah Md Toufiqur Rahman, Mohammad Aqdas, Erik W Martin, Francesco Tomassoni Ardori, Preeyaporn Songkiatisak, Kyu-Seon Oh, Stefan Uderhardt, Sangwon Yun, Quia C Claybourne, Ross A McDevitt, Valentina Greco, Ronald N Germain, Lino Tessarollo, Myong-Hee Sung
ABSTRACT
In vitro studies suggest that mapping the spatiotemporal complexity of nuclear factor κB (NF-κB) signaling is essential to understanding its function. The lack of tools to directly monitor NF-κB proteins in vivo has hindered such efforts. Here, we introduce reporter mice with the endogenous RelA (p65) or c-Rel labeled with distinct fluorescent proteins and a double knockin with both subunits labeled. Overcoming hurdles in simultaneous live-cell imaging of RelA and c-Rel, we show that quantitative features of signaling reflect the identity of activating ligands, differ between primary and immortalized cells, and shift toward c-Rel in microglia from aged brains. RelA:c-Rel heterodimer is unexpectedly depleted in the nuclei of stimulated cells. Trajectories of subunit co-expression in immune lineages reveal a reduction at key cell maturation stages. These results demonstrate the power of these reporters in gaining deeper insights into NF-κB biology, with the spectral complementarity of the labeled NF-κB proteins enabling diverse applications.
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Cycloheximide, ≥90% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Goat serum