Merck
  • Gpr37l1/prosaposin receptor regulates Ptch1 trafficking, Shh production, and cell proliferation in cerebellar primary astrocytes.
Gpr37l1/prosaposin receptor regulates Ptch1 trafficking, Shh production, and cell proliferation in cerebellar primary astrocytes.
Journal of neuroscience research (2020-12-23)
Gina La Sala, Chiara Di Pietro, Rafaele Matteoni, Giulia Bolasco, Daniela Marazziti, Glauco P Tocchini-Valentini
ABSTRACT
Mammalian cerebellar astrocytes critically regulate the differentiation and maturation of neuronal Purkinje cells and granule precursors. The G protein-coupled receptor 37-like 1 (Gpr37l1) is expressed by Bergmann astrocytes and interacts with patched 1 (Ptch1) at peri-ciliary membranes. Cerebellar primary astrocyte cultures from wild-type and Gpr37l1 null mutant mouse pups were established and studied. Primary cilia were produced by cultures of both genotypes, as well as Ptch1 and smoothened (Smo) components of the sonic hedgehog (Shh) mitogenic pathway. Compared to wild-type cells, Gpr37l1-/- astrocytes displayed striking increases in proliferative activity, Ptch1 protein expression and internalization, intracellular cholesterol content, ciliary localization of Smo, as well as a marked production of active Shh. Similar effects were reproduced by treating wild-type astrocytes with a putative prosaptide ligand of Gpr37l1. These findings indicate that Gpr37l1-Ptch1 interactions specifically regulate Ptch1 internalization and trafficking, with consequent stimulation of Shh production and activation of proliferative signaling.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-JMJD1A Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
SANT-1, A potent, cell-permeable antagonist of the Shh signaling pathway by binding directly to Smoothened, a distant relative of G protein-coupled receptors.
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, clone DM1A, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, Alexa Fluor488 Conjugate | MABN50A4, clone 211F1.1, from mouse, ALEXA FLUOR 488