- Stimulatory effects of platycodin D on osteoblast differentiation.
Stimulatory effects of platycodin D on osteoblast differentiation.
Previous studies have suggested that platycodin D is implicated in bone biology and ameliorates osteoporosis development. Platycodin D repressed the osteoclast activity and enhanced bone mineral density in the mouse model. However, the effects of platycodin D on osteoblast differentiation have not been elucidated yet. In C3H10T1/2 cells, platycodin D upregulated osteogenic markers including alkaline phosphatase (ALP), bone sialoprotein, and collagen type 1 alpha 1, and transcription factors, such as Runx2 and osterix, subsequently enhancing the bone mineralization. In a molecular mechanism study, platycodin D induced β-catenin nuclear accumulation by upregulating GSK3β phosphorylation. Furthermore, platycodin D upregulated the ALP activity and enhanced the mineralization process in osteoblast cells via the sirtuin 1/β-catenin pathways. Taken together, these results suggested that platycodin D could be an effective therapeutic compound against osteoporosis because of its regulatory effects during the osteoblast differentiation.