Merck
  • The Grb2 splice variant, Grb3-3, is a negative regulator of RAS activation.
The Grb2 splice variant, Grb3-3, is a negative regulator of RAS activation.
Communications biology (2022-09-29)
Caroline Seiler, Amy K Stainthorp, Sophie Ketchen, Christopher M Jones, Kate Marks, Philip Quirke, John E Ladbury
ABSTRACT
Activation of RAS is crucial in driving cellular outcomes including proliferation, differentiation, migration and apoptosis via the MAPK pathway. This is initiated on recruitment of Grb2, as part of a Grb2-Sos complex, to an up-regulated receptor tyrosine kinase (RTK), enabling subsequent interaction of Sos with the plasma membrane-localised RAS. Aberrant regulation at this convergence point for RTKs in MAPK signalling is a key driver of multiple cancers. Splicing of the GRB2 gene produces a deletion variant, Grb3-3, that is incapable of binding to RTKs. We show that, despite maintaining the ability to bind to Sos, the Grb3-3-Sos complex remains in the cytoplasm, unable to engage with RAS. Competition between Grb2 and Grb3-3 for binding to C-terminal proline-rich sequences on Sos modulates MAPK signalling. Additionally, we demonstrate that splicing is regulated by heterogenous nuclear riboproteins C1/C2, and that normal and malignant colon tissue show differential Grb3-3 expression.
Sigma-Aldrich
Monoclonal Anti-SOS1 antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%