Merck
  • Effector membrane translocation biosensors reveal G protein and βarrestin coupling profiles of 100 therapeutically relevant GPCRs.
Effector membrane translocation biosensors reveal G protein and βarrestin coupling profiles of 100 therapeutically relevant GPCRs.
eLife (2022-03-19)
Charlotte Avet, Arturo Mancini, Billy Breton, Christian Le Gouill, Alexander S Hauser, Claire Normand, Hiroyuki Kobayashi, Florence Gross, Mireille Hogue, Viktoriya Lukasheva, Stéphane St-Onge, Marilyn Carrier, Madeleine Héroux, Sandra Morissette, Eric B Fauman, Jean-Philippe Fortin, Stephan Schann, Xavier Leroy, David E Gloriam, Michel Bouvier
ABSTRACT
The recognition that individual GPCRs can activate multiple signaling pathways has raised the possibility of developing drugs selectively targeting therapeutically relevant ones. This requires tools to determine which G proteins and βarrestins are activated by a given receptor. Here, we present a set of BRET sensors monitoring the activation of the 12 G protein subtypes based on the translocation of their effectors to the plasma membrane (EMTA). Unlike most of the existing detection systems, EMTA does not require modification of receptors or G proteins (except for Gs). EMTA was found to be suitable for the detection of constitutive activity, inverse agonism, biased signaling and polypharmacology. Profiling of 100 therapeutically relevant human GPCRs resulted in 1500 pathway-specific concentration-response curves and revealed a great diversity of coupling profiles ranging from exquisite selectivity to broad promiscuity. Overall, this work describes unique resources for studying the complexities underlying GPCR signaling and pharmacology.
Sigma-Aldrich
Probenecid
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Parathyroid Hormone Fragment 1-34 human, ≥95% (HPLC), powder
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
Anti-GNA11 (C-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Lithocholic acid, ≥95%
Sigma-Aldrich
α-Melanocyte stimulating hormone, ≥97% (HPLC)
Sigma-Aldrich
[Arg8]-Vasopressin acetate salt, ≥95% (HPLC)