Merck
  • Synthesis of breast cancer targeting conjugate of temporin-SHa analog and its effect on pro- and anti-apoptotic protein expression in MCF-7 cells.
Synthesis of breast cancer targeting conjugate of temporin-SHa analog and its effect on pro- and anti-apoptotic protein expression in MCF-7 cells.
Peptides (2018-07-22)
Farzana Shaheen, Muhammad Nadeem-Ul-Haque, Aqeel Ahmed, Shabana U Simjee, A Ganesan, Almas Jabeen, Zafar Ali Shah, M Iqbal Choudhary
ABSTRACT
The frog natural product temporin-SHa (FLSGIVGMLGKLFamide) is a potent antimicrobial peptide, as is the analog [S3K]SHa. By solid-phase synthesis, we prepared temporin-SHa and several temporin-SHa analogs with one or more D-alanine residues incorporated. The natural product and the analog [G10a]SHa were found to be cytotoxic in mammalian cell lines and induce cell death. To achieve selectivity, we conjugated the analog [G10a]SHa with a breast cancer targeting peptide (BCTP). The resulting peptide temporin [G10a]SHa-BCTP conjugate was selectively active against the MCF-7 breast cancer cell line with no cytotoxicity in NIH-3T3 fibroblasts. Unlike the natural product or [G10a]SHa, the conjugated peptide induced apoptosis, downregulating the expression of Bcl-2 and survivin and upregulating Bax and caspase-3.
Sigma-Aldrich
Apo-Direct TUNEL Assay Kit, The APO-DIRECT Kit is a single-step staining method for labeling DNA breaks to detect apoptotic cells by flow cytometry.