Merck
  • Cytosolic localization and in vitro assembly of human de novo thymidylate synthesis complex.
Cytosolic localization and in vitro assembly of human de novo thymidylate synthesis complex.
The FEBS journal (2021-10-26)
Sharon Spizzichino, Dalila Boi, Giovanna Boumis, Roberta Lucchi, Francesca Romana Liberati, Davide Capelli, Roberta Montanari, Giorgio Pochetti, Roberta Piacentini, Giacomo Parisi, Alessio Paone, Serena Rinaldo, Roberto Contestabile, Angela Tramonti, Alessandro Paiardini, Giorgio Giardina, Francesca Cutruzzolà
ABSTRACT
De novo thymidylate synthesis is a crucial pathway for normal and cancer cells. Deoxythymidine monophosphate (dTMP) is synthesized by the combined action of three enzymes: serine hydroxymethyltransferase (SHMT1), dihydrofolate reductase (DHFR) and thymidylate synthase (TYMS), with the latter two being targets of widely used chemotherapeutics such as antifolates and 5-fluorouracil. These proteins translocate to the nucleus after SUMOylation and are suggested to assemble in this compartment into the thymidylate synthesis complex. We report the intracellular dynamics of the complex in cancer cells by an in situ proximity ligation assay, showing that it is also detected in the cytoplasm. This result indicates that the role of the thymidylate synthesis complex assembly may go beyond dTMP synthesis. We have successfully assembled the dTMP synthesis complex in vitro, employing tetrameric SHMT1 and a bifunctional chimeric enzyme comprising human thymidylate synthase and dihydrofolate reductase. We show that the SHMT1 tetrameric state is required for efficient complex assembly, indicating that this aggregation state is evolutionarily selected in eukaryotes to optimize protein-protein interactions. Lastly, our results regarding the activity of the complete thymidylate cycle in vitro may provide a useful tool with respect to developing drugs targeting the entire complex instead of the individual components.
Sigma-Aldrich
Monoclonal Anti-DHFR antibody produced in mouse, clone 2B10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Anti-α-Tubulin−FITC antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2