Merck
  • Chronic intake of high dietary sucrose induces sexually dimorphic metabolic adaptations in mouse liver and adipose tissue.
Chronic intake of high dietary sucrose induces sexually dimorphic metabolic adaptations in mouse liver and adipose tissue.
Nature communications (2022-10-14)
Erin J Stephenson, Amanda S Stayton, Aarti Sethuraman, Prahlad K Rao, Alice Meyer, Charles Klazer Gomes, Molly C Mulcahy, Liam McAllan, Michelle A Puchowicz, Joseph F Pierre, Dave Bridges, Joan C Han
ABSTRACT
Almost all effective treatments for non-alcoholic fatty liver disease (NAFLD) involve reduction of adiposity, which suggests the metabolic axis between liver and adipose tissue is essential to NAFLD development. Since excessive dietary sugar intake may be an initiating factor for NAFLD, we have characterized the metabolic effects of liquid sucrose intake at concentrations relevant to typical human consumption in mice. We report that sucrose intake induces sexually dimorphic effects in liver, adipose tissue, and the microbiome; differences concordant with steatosis severity. We show that when steatosis is decoupled from impairments in insulin responsiveness, sex is a moderating factor that influences sucrose-driven lipid storage and the contribution of de novo fatty acid synthesis to the overall hepatic triglyceride pool. Our findings provide physiologic insight into how sex influences the regulation of adipose-liver crosstalk and highlight the importance of extrahepatic metabolism in the pathogenesis of diet-induced steatosis and NAFLD.
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Anti-PKLR antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Serum Triglyceride Determination Kit, 1 kit sufficient for 250 tests