Merck
  • Isolation of mouse pancreatic islet Procr+ progenitors and long-term expansion of islet organoids in vitro.
Isolation of mouse pancreatic islet Procr+ progenitors and long-term expansion of islet organoids in vitro.
Nature protocols (2022-04-10)
Jingqiang Wang, Daisong Wang, Xinyi Chen, Shubo Yuan, Lanyue Bai, Chunye Liu, Yi Arial Zeng
ABSTRACT
Insulin production is required for glucose homeostasis. Pancreatic islet β cells are the only cells that produce insulin in humans; however, generation of functional β cells in vitro from embryonic or adult tissues has been challenging. Here, we describe isolation of pancreatic islet progenitors from adult mice, which enables the efficient generation and long-term expansion of functional islet organoids in vitro. This protocol starts with purification of protein C receptor (Procr)-expressing islet progenitors. Coculture with endothelial cells generates islet organoids in vitro that can be expanded by passage. Functional maturation is achieved as a consequence of a prolonged culture period and cyclic glucose stimulation. Primary islet organoids form in 7-10 days. Subsequently, each passage takes 1 week, with the final maturation step requiring 3 weeks of additional culture. The resulting organoids are predominantly composed of β cells but also contain small proportions of α, δ and pancreatic polypeptide cells. The organoids sense glucose and secrete insulin. This approach thus provides a strategy for β cell generation in vitro and an organoid system to study islet regeneration and diseases.
Sigma-Aldrich
Anti-Pancreatic polypeptide/PPY antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat, affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-SST antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution