Merck
  • Stimulated emission depletion microscopy with a single depletion laser using five fluorochromes and fluorescence lifetime phasor separation.
Stimulated emission depletion microscopy with a single depletion laser using five fluorochromes and fluorescence lifetime phasor separation.
Scientific reports (2022-08-19)
Mariano Gonzalez Pisfil, Iliya Nadelson, Brigitte Bergner, Sonja Rottmeier, Andreas W Thomae, Steffen Dietzel
ABSTRACT
Stimulated emission depletion (STED) microscopy achieves super-resolution by exciting a diffraction-limited volume and then suppressing fluorescence in its outer parts by depletion. Multiple depletion lasers may introduce misalignment and bleaching. Hence, a single depletion wavelength is preferable for multi-color analyses. However, this limits the number of usable spectral channels. Using cultured cells, common staining protocols, and commercially available fluorochromes and microscopes we exploit that the number of fluorochromes in STED or confocal microscopy can be increased by phasor based fluorescence lifetime separation of two dyes with similar emission spectra but different fluorescent lifetimes. In our multi-color FLIM-STED approach two fluorochromes in the near red (exc. 594 nm, em. 600-630) and two in the far red channel (633/641-680), supplemented by a single further redshifted fluorochrome (670/701-750) were all depleted with a single laser at 775 nm thus avoiding potential alignment issues. Generally, this approach doubles the number of fully distinguishable colors in laser scanning microscopy. We provide evidence that eight color FLIM-STED with a single depletion laser would be possible if suitable fluorochromes were identified and we confirm that a fluorochrome may have different lifetimes depending on the molecules to which it is coupled.
Sigma-Aldrich
Anti-TOMM20 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross-adsorbed, CF 680R antibody produced in donkey, ~2 mg/mL, affinity isolated antibody