Merck
  • Distinct spatio-temporal extracellular matrix accumulation within demyelinated spinal cord lesions in Theiler's murine encephalomyelitis.
Distinct spatio-temporal extracellular matrix accumulation within demyelinated spinal cord lesions in Theiler's murine encephalomyelitis.
Brain pathology (Zurich, Switzerland) (2011-07-20)
Verena Haist, Reiner Ulrich, Arno Kalkuhl, Ulrich Deschl, Wolfgang Baumgärtner
ABSTRACT
The accumulation of extracellular matrix (ECM) and glial scar formation are considered important factors for the failure of regeneration in central nervous system (CNS) injury and multiple sclerosis. Theiler's murine encephalomyelitis (TME) as a model of multiple sclerosis served to evaluate the spatio-temporal course of ECM alterations in demyelinating conditions. Microarray analysis revealed only mildly upregulated gene expression of ECM molecules, their biosynthesis pathways and pro-fibrotic factors, while upregulation of matrix remodeling enzymes was more prominent. Immunohistochemistry demonstrated progressive accumulation of chondroitin sulfate proteoglycans, glycoproteins and collagens within demyelinated TME lesions, paralleling the development of astrogliosis. Deposition of collagen IV, laminin, perlecan and tenascin-C started 28 days postinfection (dpi), collagen I, decorin, entactin and neurocan accumulated from 56 dpi on, and fibronectin from 98 dpi on. The basement membrane (BM) molecules collagen IV, entactin, fibronectin, laminin and perlecan showed perivascular and parenchymal deposition, while the non-BM components collagen I, decorin, neurocan and tenascin-C only accumulated in a nonvascular pattern in demyelinated areas. Contrary, phosphacan expression progressively decreased during TME. The immunoreactivity of aggrecan and brevican remained unchanged. The spatio-temporal association of matrix accumulation with astrogliosis suggests a mainly astrocytic origin of ECM deposits, which in turn may contribute to remyelination failure in TME.
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Phosphacan Antibody, clone 122.2, ascites fluid, clone 122.2, Chemicon®
Sigma-Aldrich
Anti-Neurocan Antibody, clone 650.24, clone 650.24, Chemicon®, from mouse
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution