Merck
  • The BTB transcription factors ZBTB11 and ZFP131 maintain pluripotency by repressing pro-differentiation genes.
The BTB transcription factors ZBTB11 and ZFP131 maintain pluripotency by repressing pro-differentiation genes.
Cell reports (2022-03-17)
Görkem Garipler, Congyi Lu, Alexis Morrissey, Lorena S Lopez-Zepeda, Yingzhen Pei, Simon E Vidal, Ana Paula Zen Petisco Fiore, Begüm Aydin, Matthias Stadtfeld, Uwe Ohler, Shaun Mahony, Neville E Sanjana, Esteban O Mazzoni
ABSTRACT
In pluripotent cells, a delicate activation-repression balance maintains pro-differentiation genes ready for rapid activation. The identity of transcription factors (TFs) that specifically repress pro-differentiation genes remains obscure. By targeting ∼1,700 TFs with CRISPR loss-of-function screen, we found that ZBTB11 and ZFP131 are required for embryonic stem cell (ESC) pluripotency. ESCs without ZBTB11 or ZFP131 lose colony morphology, reduce proliferation rate, and upregulate transcription of genes associated with three germ layers. ZBTB11 and ZFP131 bind proximally to pro-differentiation genes. ZBTB11 or ZFP131 loss leads to an increase in H3K4me3, negative elongation factor (NELF) complex release, and concomitant transcription at associated genes. Together, our results suggest that ZBTB11 and ZFP131 maintain pluripotency by preventing premature expression of pro-differentiation genes and present a generalizable framework to maintain cellular potency.
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Sigma-Aldrich
Anti-NELFE antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution