Merck
  • Growth arrest of PPP2R5C and PPP2R5D double knockout mice indicates a genetic interaction and conserved function for these PP2A B subunits.
Growth arrest of PPP2R5C and PPP2R5D double knockout mice indicates a genetic interaction and conserved function for these PP2A B subunits.
FASEB bioAdvances (2022-04-14)
Jade J Dyson, Fatima Abbasi, Prajakta Varadkar, Brent McCright
ABSTRACT
Protein phosphatase 2A (PP2A) is a heterotrimeric phosphatase that controls a wide range of cellular functions. The catalytic activity and intracellular location of PP2A are modulated by its association with regulatory B subunits, including B56 proteins, which are encoded by five separate genes in humans and mice. The specific effects of each B56 protein on PP2A activity and function are largely unknown. As part of an effort to identify specific PP2A-B56 functions, we created knockout strains of B56β, B56δ, and B56ε using CRISPR/Cas9n. We found that none of the individual B56 genes are essential for mouse survival. However, mice that have both B56δ and B56γ inactivated (B56δγ-), arrest fetal development around Day E12. The hearts of B56δγ- mice have a single outflow vessel rather than having both an aorta and a pulmonary artery. Thus, there appears to be strong genetic interaction between B56δ and B56γ, and together they are necessary for heart development. Of note, both these proteins have been shown to localize to the nucleus and have the most related peptide sequences of the B56 family members. Our results suggest there are B56 subfamilies, which work in conjunction to regulate specific PP2A functions.
Sigma-Aldrich
Anti-PPP2R5D antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
Sigma-Aldrich
Anti-PPP2R5B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-PP2A reg. subunit B56 epsilon (PPP2R5E) Antibody, clone 5A5-1F3, clone 5A5-1F3, from mouse