Merck
  • Aberrant macrophage cytokine production is a conserved feature among autoimmune-prone mouse strains: elevated interleukin (IL)-12 and an imbalance in tumor necrosis factor-alpha and IL-10 define a unique cytokine profile in macrophages from young nonobese diabetic mice.
Aberrant macrophage cytokine production is a conserved feature among autoimmune-prone mouse strains: elevated interleukin (IL)-12 and an imbalance in tumor necrosis factor-alpha and IL-10 define a unique cytokine profile in macrophages from young nonobese diabetic mice.
Diabetes (2000-07-26)
D G Alleva, R P Pavlovich, C Grant, S B Kaser, D I Beller
ABSTRACT
Cytokines derived from macrophages (Mø) play a critical role in the development of type 1 diabetes in the nonobese diabetic (NOD) mouse. Based on earlier findings from lupus-prone strains of inherent cytokine defects in Mø , NOD Mø were evaluated for intrinsically dysregulated cytokine production with the potential to initiate or exacerbate disease. Endotoxin-activated peritoneal Mø from young prediseased NOD mice produced interleukin (IL)-1 and tumor necrosis factor (TNF)-alpha levels similar to those of Mø from a panel of control strains but reduced compared with the congenic diabetes-resistant NOR strain. IL-6 and IL-10 production were similar in NOD and NOR Mø, indicating that reduction in NOD IL-1 and TNF-alpha expression was selective. Nevertheless, the ratio of TNF-alpha and IL-10 production, a stringent index of normal Mø function, distinguished NOD from all normal strains. The most striking feature of NOD Mø, however, was their substantially elevated IL-12 production. This response was induced not only by endotoxin but also by bacillus Calmette-Guerin (BCG) and CD40 ligand and was associated with (and likely caused by) the enhanced and prolonged expression of p40 mRNA. Moreover, NOD Mø IL-12 expression appeared to be near maximally induced by lipopolysaccharide (LPS) alone, because it was only slightly enhanced by the addition of gamma-interferon, a stimulus that substantially elevated LPS-induced IL-12 production in Mø from normal strains. Accompanied by a unique profile of TNF-alpha and IL-10, the dramatic elevation of IL-12 expression by NOD Mø reflects intrinsic defects of the innate immune system with the potential to initiate and propagate the pathogenic autoreactive T-helper type 1 response characteristic of type 1 diabetes.
Sigma-Aldrich
Avidin–Peroxidase, lyophilized powder