Merck
  • Multiple LEDT wavelengths modulate the Akt signaling pathways and attenuate pathological events in mdx dystrophic muscle cells.
Multiple LEDT wavelengths modulate the Akt signaling pathways and attenuate pathological events in mdx dystrophic muscle cells.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2022-04-06)
Guilherme Luiz da Rocha, Daniela Sayuri Mizobuti, Heloina Nathalliê Mariano da Silva, Caroline Covatti, Caroline Caramano de Lourenço, Marcos José Salvador, Elaine Cristina Leite Pereira, Elaine Minatel
ABSTRACT
This study is aimed at investigating the effects of LEDT, at multiple wavelengths, on intracellular calcium concentration; on transient receptor potential canonical channels; on calcium-binding protein; on myogenic factors; on myosin heavy chains; on Akt signaling pathway; on inflammatory markers; and on the angiogenic-inducing factor in dystrophic muscle cell culture experimental model. Dystrophic primary muscle cells were submitted to LEDT, at multiple wavelengths (420 nm, 470 nm, 660 nm, and 850 nm), and evaluated after 48 h for cytotoxic effects and intracellular calcium content. TRPC-1, TRPC-6, Calsequestrin, MyoD, Myogenin, MHC-slow, MHC-fast, p-AKT, p-mTOR, p-FoxO1, Myostatin, NF-κB, TNF-α, and VEGF levels were evaluated in dystrophic primary muscle cells by western blotting. The LEDT, at multiple wavelengths, treated-mdx muscle cells showed no cytotoxic effect and significant lower levels in [Ca2 +]i. The mdx muscle cells treated with LEDT showed a significant reduction of TRPC-1, NF-κB, TNF-α and MyoD levels and a significant increase of Myogenin, MHC-slow, p-AKT, p-mTOR, p-FoxO1 levels, and VEGF levels. Our findings suggest that different LEDT wavelengths modulate the Akt-signaling pathways and attenuate pathological events in dystrophic muscle cells, and a combined multiwavelength irradiation protocol may even provide a potentially therapeutic strategy for muscular dystrophies.
Sigma-Aldrich
Anti-FOXO1 (Ab-256) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-Myosin (Skeletal, Fast) antibody produced in mouse, clone MY-32, ascites fluid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Myogenin antibody produced in mouse, clone F12B, purified immunoglobulin, buffered aqueous solution