Merck
  • Laminin-10/11 and fibronectin differentially regulate integrin-dependent Rho and Rac activation via p130(Cas)-CrkII-DOCK180 pathway.
Laminin-10/11 and fibronectin differentially regulate integrin-dependent Rho and Rac activation via p130(Cas)-CrkII-DOCK180 pathway.
The Journal of biological chemistry (2001-05-23)
J Gu, Y Sumida, N Sanzen, K Sekiguchi
ABSTRACT
The alpha(5) chain-containing laminin isoforms, laminins-10 and -11 (laminin-10/11), are the major components of the basement membrane, having potent cell-adhesive activity. We examined the cell-adhesive and integrin-mediated signaling activities of laminin-10/11 in comparison to fibronectin, the best characterized extracellular adhesive ligand. We found that laminin-10/11 are more active than fibronectin in promoting cell migration and preferentially activate Rac, not Rho, via the p130(Cas)-CrkII-DOCK180 pathway. Cells adhering to fibronectin develop stress fibers and focal contacts, whereas cells adhering to laminin-10/11 do not, consistent with the high cell migration-promoting activity of laminin-10/11. Pull-down assays of GTP-loaded Rac and Rho demonstrated the preferential activation of Rac on laminin-10/11, in contrast to the activation of Rho on fibronectin. Activation of Rac by laminin-10/11 was associated with the phosphorylation of p130(Cas) and an increased formation of a p130(Cas)-CrkII-DOCK180 complex. Cell migration on laminin-10/11 was suppressed by the expression of either a dominant-negative Rac or CrkII mutants defective in p130(Cas) or DOCK180 binding. This is the first report demonstrating a distinct activation of Rho family GTPases resulting from adhesion to different extracellular ligands.
Sigma-Aldrich
Anti-Integrin β4 Antibody, clone ASC-8, clone ASC-8, Chemicon®, from mouse