Merck
  • Muscle-derived fibro-adipogenic progenitor cells for production of cultured bovine adipose tissue.
Muscle-derived fibro-adipogenic progenitor cells for production of cultured bovine adipose tissue.
NPJ science of food (2022-01-26)
Richard G J Dohmen, Sophie Hubalek, Johanna Melke, Tobias Messmer, Federica Cantoni, Arianna Mei, Rui Hueber, Rada Mitic, Dirk Remmers, Panagiota Moutsatsou, Mark J Post, Laura Jackisch, Joshua E Flack
ABSTRACT
Cultured meat is an emergent technology with the potential for significant environmental and animal welfare benefits. Accurate mimicry of traditional meat requires fat tissue; a key contributor to both the flavour and texture of meat. Here, we show that fibro-adipogenic progenitor cells (FAPs) are present in bovine muscle, and are transcriptionally and immunophenotypically distinct from satellite cells. These two cell types can be purified from a single muscle sample using a simple fluorescence-activated cell sorting (FACS) strategy. FAPs demonstrate high levels of adipogenic potential, as measured by gene expression changes and lipid accumulation, and can be proliferated for a large number of population doublings, demonstrating their suitability for a scalable cultured meat production process. Crucially, FAPs reach a mature level of adipogenic differentiation in three-dimensional, edible hydrogels. The resultant tissue accurately mimics traditional beef fat in terms of lipid profile and taste, and FAPs thus represent a promising candidate cell type for the production of cultured fat.
Sigma-Aldrich
Monoclonal Anti-Desmin antibody produced in mouse, clone DE-U-10, ascites fluid
Sigma-Aldrich
Collagen solution from bovine skin, 6 mg/mL, sterile-filtered, BioReagent, suitable for cell culture