Merck
Subcellular mechanisms of endothelin action in vascular system.
European journal of pharmacology (1999-08-12)
T Masaki, S Miwa, T Sawamura, H Ninomiya, Y Okamoto
ABSTRACT
To elucidate the role of endothelin in the regulation of vascular function, the cellular and subcellular mechanisms for the synthesis of endothelin and the function of endothelin-receptors have been studied extensively. In this article, recent results regarding these problems are reviewed. (1) Oxidatively modified low-density-lipoprotein (LDL) reduces nitric oxide (NO) release via inhibition of the high-affinity arginine transporter of endothelial cells. (2) Endothelin-1-induced vasoconstriction is mediated by Ca2+ influx through a non-selective cation channel sensitive to 1-[beta-[3-(4-methoxyphenyl) propoxyl]-4-methoxyphenethyl]-1H-imidazole HCl (SK & F96365). (3) A distinct domain of the endothelin-receptor is required for the coupling of different G(alpha)-proteins. (4) Endothelin ET(A) receptor-mediated mitogenic activity is mediated by two pathways, one classical protein kinase C(PKC)-dependent, and the other phosphoinositide 3-kinase dependent. Both stimulate mitogen-activated protein kinase (MAPK). Endothelin ET(B) receptor-mediated mitogenic activity is also mediated by the PKC-dependent pathway. In contrast, endothelin ET(B) receptor-mediates differentiation and apoptosis via G(alpha)i coupling.
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder