Merck
  • RhoA/Cdc42 signaling drives cytoplasmic maturation but not endomitosis in megakaryocytes.
RhoA/Cdc42 signaling drives cytoplasmic maturation but not endomitosis in megakaryocytes.
Cell reports (2021-05-13)
Tobias Heib, Heike M Hermanns, Georgi Manukjan, Maximilian Englert, Charly Kusch, Isabelle Carlotta Becker, Annika Gerber, Lou Martha Wackerbarth, Philipp Burkard, Thomas Dandekar, Johannes Balkenhol, Daniel Jahn, Sarah Beck, Mara Meub, Sebastian Dütting, Christian Stigloher, Markus Sauer, Deya Cherpokova, Harald Schulze, Cord Brakebusch, Bernhard Nieswandt, Zoltan Nagy, Irina Pleines
ABSTRACT
Megakaryocytes (MKs), the precursors of blood platelets, are large, polyploid cells residing mainly in the bone marrow. We have previously shown that balanced signaling of the Rho GTPases RhoA and Cdc42 is critical for correct MK localization at bone marrow sinusoids in vivo. Using conditional RhoA/Cdc42 double-knockout (DKO) mice, we reveal here that RhoA/Cdc42 signaling is dispensable for the process of polyploidization in MKs but essential for cytoplasmic MK maturation. Proplatelet formation is virtually abrogated in the absence of RhoA/Cdc42 and leads to severe macrothrombocytopenia in DKO animals. The MK maturation defect is associated with downregulation of myosin light chain 2 (MLC2) and β1-tubulin, as well as an upregulation of LIM kinase 1 and cofilin-1 at both the mRNA and protein level and can be linked to impaired MKL1/SRF signaling. Our findings demonstrate that MK endomitosis and cytoplasmic maturation are separately regulated processes, and the latter is critically controlled by RhoA/Cdc42.
Sigma-Aldrich
LIM Kinase Inhibitor I, LIMKi 3, The LIM Kinase Inhibitor I, LIMKi 3 controls the biological activity of LIM Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Anti-Tubulin Antibody, clone YL1/2, clone YL1/2, Chemicon®, from rat
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder