Merck
WASH interacts with Ku to regulate DNA double-stranded break repair.
iScience (2022-01-18)
Tao Wang, Xiao-Hui Du, Yu Hong, Xian Hong, Li Fan, Jian-Wen Zhou, He Sun, Jie Ge, Daniel D Billadeau, Zhi-Hui Deng
ABSTRACT
The Wiskott-Aldrich syndrome protein and SCAR homolog (WASH), an actin nucleation-promoting factor, is present in the nucleus where it regulates gene transcription and maintains nuclear organization. Here, we show that WASH interacts with core non-homologous end-joining (NHEJ) factors including Ku70/Ku80 and DNA-PKcs, and Ku70/Ku80 is involved in the recruitment of WASH to the sites of DNA double-stranded break (DSB). WASH depletion leads to increased cell sensitivity and impaired DNA repair capacity in response to etoposide-induced DSBs and reduces NHEJ efficiency. Mechanistically, we show that loss of WASH inhibits the phosphorylation of DNA-PKcs, H2AX, and KAP1 after DSB induction and reduces chromatin relaxation and the recruitment of several downstream NHEJ factors to DSBs. Moreover, WASH role in DSB repair depends on its conserved C-terminal VCA domain and Arp2/3 activation. Our findings reveal a function and mechanistic insight for WASH in DNA DSB repair by the NHEJ pathway.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Millipore
EZview Red Anti-HA Affinity Gel
Millipore
Protein A (extracellular)–Agarose from Staphylococcus aureus, lyophilized powder
Millipore
Protein G–Agarose, lyophilized powder, Contains lactose stabilizers that must be removed prior to use.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)