Merck
  • Orai1α, but not Orai1β, co-localizes with TRPC1 and is required for its plasma membrane location and activation in HeLa cells.
Orai1α, but not Orai1β, co-localizes with TRPC1 and is required for its plasma membrane location and activation in HeLa cells.
Cellular and molecular life sciences : CMLS (2022-01-07)
Jose Sanchez-Collado, Jose J Lopez, Isaac Jardin, Alejandro Berna-Erro, Pedro J Camello, Carlos Cantonero, Tarik Smani, Gines M Salido, Juan A Rosado
ABSTRACT
The identification of two variants of the canonical pore-forming subunit of the Ca2+ release-activated Ca2+ (CRAC) channel Orai1, Orai1α and Orai1β, in mammalian cells arises the question whether they exhibit different functional characteristics. Orai1α and Orai1β differ in the N-terminal 63 amino acids, exclusive of Orai1α, and show different sensitivities to Ca2+-dependent inactivation, as well as distinct ability to form arachidonate-regulated channels. We have evaluated the role of both Orai1 variants in the activation of TRPC1 in HeLa cells. We found that Orai1α and Orai1β are required for the maintenance of regenerative Ca2+ oscillations, while TRPC1 plays a role in agonist-induced Ca2+ influx but is not essential for Ca2+ oscillations. Using APEX2 proximity labeling, co-immunoprecipitation and the fluorescence of G-GECO1.2 fused to Orai1α our results indicate that agonist stimulation and Ca2+ store depletion enhance Orai1α-TRPC1 interaction. Orai1α is essential for TRPC1 plasma membrane location and activation. Thus, TRPC1 function in HeLa cells depends on Ca2+ influx through Orai1α exclusively.
Sigma-Aldrich
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-ORAI1 (AB1) antibody produced in rabbit, affinity isolated antibody
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution