Merck
  • MAP1B regulates axonal development by modulating Rho-GTPase Rac1 activity.
MAP1B regulates axonal development by modulating Rho-GTPase Rac1 activity.
Molecular biology of the cell (2010-08-20)
Carolina Montenegro-Venegas, Elena Tortosa, Silvana Rosso, Diego Peretti, Flavia Bollati, Mariano Bisbal, Ignacio Jausoro, Jesus Avila, Alfredo Cáceres, Christian Gonzalez-Billault
ABSTRACT
Cultured neurons obtained from MAP1B-deficient mice have a delay in axon outgrowth and a reduced rate of axonal elongation compared with neurons from wild-type mice. Here we show that MAP1B deficiency results in a significant decrease in Rac1 and cdc42 activity and a significant increase in Rho activity. We found that MAP1B interacted with Tiam1, a guanosine nucleotide exchange factor for Rac1. The decrease in Rac1/cdc42 activity was paralleled by decreases in the phosphorylation of the downstream effectors of these proteins, such as LIMK-1 and cofilin. The expression of a constitutively active form of Rac1, cdc42, or Tiam1 rescued the axon growth defect of MAP1B-deficient neurons. Taken together, these observations define a new and crucial function of MAP1B that we show to be required for efficient cross-talk between microtubules and the actin cytoskeleton during neuronal polarization.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid