Merck
  • Reduced NCOR2 expression accelerates androgen deprivation therapy failure in prostate cancer.
Reduced NCOR2 expression accelerates androgen deprivation therapy failure in prostate cancer.
Cell reports (2021-12-16)
Mark D Long, Justine J Jacobi, Prashant K Singh, Gerard Llimos, Sajad A Wani, Aryn M Rowsam, Spencer R Rosario, Marlous Hoogstraat, Simon Linder, Jason Kirk, Hayley C Affronti, Andries Bergman, Wilbert Zwart, Moray J Campbell, Dominic J Smiraglia
ABSTRACT
This study addresses the roles of nuclear receptor corepressor 2 (NCOR2) in prostate cancer (PC) progression in response to androgen deprivation therapy (ADT). Reduced NCOR2 expression significantly associates with shorter disease-free survival in patients with PC receiving adjuvant ADT. Utilizing the CWR22 xenograft model, we demonstrate that stably reduced NCOR2 expression accelerates disease recurrence following ADT, associates with gene expression patterns that include neuroendocrine features, and induces DNA hypermethylation. Stably reduced NCOR2 expression in isogenic LNCaP (androgen-sensitive) and LNCaP-C4-2 (androgen-independent) cells revealed that NCOR2 reduction phenocopies the impact of androgen treatment and induces global DNA hypermethylation patterns. NCOR2 genomic binding is greatest in LNCaP-C4-2 cells and most clearly associates with forkhead box (FOX) transcription factor FOXA1 binding. NCOR2 binding significantly associates with transcriptional regulation most when in active enhancer regions. These studies reveal robust roles for NCOR2 in regulating the PC transcriptome and epigenome and underscore recent mutational studies linking NCOR2 loss of function to PC disease progression.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Androgen Receptor Antibody, from rabbit, purified by affinity chromatography