- Interleukin-2 in the development and control of inflammatory disease.
Interleukin-2 in the development and control of inflammatory disease.
Immunological reviews (2009-01-24)
Katrina K Hoyer, Hans Dooms, Luke Barron, Abul K Abbas
PMID19161413
ABSTRACT
Interleukin-2 (IL-2) has multiple, sometimes opposing, functions during an inflammatory response. It is a potent inducer of T-cell proliferation and T-helper 1 (Th1) and Th2 effector T-cell differentiation and provides T cells with a long-lasting competitive advantage resulting in the optimal survival and function of memory cells. In a regulatory role, IL-2 is important for the development, survival, and function of regulatory T cells, it enhances Fas-mediated activation-induced cell death, and it inhibits the development of inflammatory Th17 cells. Thus, in its dual and contrasting functions, IL-2 contributes to both the induction and the termination of inflammatory immune responses.