- Western blotting: sample preparation to detection.
Western blotting: sample preparation to detection.
Journal of visualized experiments : JoVE (2010-12-30)
Anna Eslami, Jesse Lujan
PMID21189462
ABSTRACT
Western blotting is an analytical technique used to detect specific proteins in a given sample of tissue homogenate or extract. It uses gel electrophoresis to separate native or denatured proteins by the length of the polypeptide (denaturing conditions) or by the 3-D structure of the protein (native/ non-denaturing conditions). The proteins are then transferred to a membrane (typically nitrocellulose or PVDF), where they are probed (detected) using antibodies specific to the target protein.