Merck
Immuno-TEM/STEM in Retinal Research.
Methods in molecular biology (Clifton, N.J.) (2018-10-17)
Sanae Sakami, Krzysztof Palczewski
ABSTRACT
Immuno-EM is a method that can determine the localization of a protein in a tissue at the ultrastructural level. Generally, membrane structures in immuno-EM specimens have very low contrast because fixation is performed without osmium. Here, by using high-angle annular dark field (HAADF)-scanning transmission electron microscopy (STEM) instead of transmission electron microscopy (TEM) for observation of immuno-EM samples, we demonstrate that photoreceptor disk membranes are clearly visible and that the procedures described in this article can be extended to visualize other membrane structures.