Merck
  • Syntaxin 4 Enrichment in β-Cells Prevents Conversion to Autoimmune Diabetes in Non-Obese Diabetic (NOD) Mice.
Syntaxin 4 Enrichment in β-Cells Prevents Conversion to Autoimmune Diabetes in Non-Obese Diabetic (NOD) Mice.
Diabetes (2021-09-25)
Eunjin Oh, Erika M McCown, Miwon Ahn, Pablo A Garcia, Sergio Branciamore, Shanshan Tang, De-Fu Zeng, Bart O Roep, Debbie C Thurmond
ABSTRACT
Syntaxin 4 (STX4), a plasma membrane-localized SNARE protein, regulates human islet β-cell insulin secretion and preservation of β-cell mass. We found that human type 1 diabetes (T1D) and NOD mouse islets show reduced β-cell STX4 expression, consistent with decreased STX4 expression, as a potential driver of T1D phenotypes. To test this hypothesis, we generated inducible β-cell-specific STX4-expressing NOD mice (NOD-iβSTX4). Of NOD-iβSTX4 mice, 73% had sustained normoglycemia vs. <20% of control NOD (NOD-Ctrl) mice by 25 weeks of age. At 12 weeks of age, before diabetes conversion, NOD-iβSTX4 mice demonstrated superior whole-body glucose tolerance and β-cell glucose responsiveness than NOD-Ctrl mice. Higher β-cell mass and reduced β-cell apoptosis were also detected in NOD-iβSTX4 pancreata compared with pancreata of NOD-Ctrl mice. Single-cell RNA sequencing revealed that islets from NOD-iβSTX4 had markedly reduced interferon-γ signaling and tumor necrosis factor-α signaling via nuclear factor-κB in islet β-cells, including reduced expression of the chemokine CCL5; CD4+ regulatory T cells were also enriched in NOD-iβSTX4 islets. These results provide a deeper mechanistic understanding of STX4 function in β-cell protection and warrant further investigation of STX4 enrichment as a strategy to reverse or prevent T1D in humans or protect β-cell grafts.
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Syntaxin 4 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Pricing and availability is not currently available.